Learn More
TARGETMOL CHEMICALS INC SALICYL-AMS 5MG
SDP
Catalog No. 502581119
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Salicyl-AMS is an inhibitor of mycobacterium auxin biosynthesis that inhibits the growth of mycobacterium in the presence of iron deficiency in vitro. purity: 99%
Catalog No. 50-258-1119 Supplier TARGETMOL CHEMICALS INC Supplier No. T171765MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Salicyl-AMS is an inhibitor of mycobacterium auxin biosynthesis that inhibits the growth of mycobacterium in the presence of iron deficiency in vitro. purity: 99%