Learn More
Frinton Laboratories SALICYLALDAZINE 25GR
Supplier:  Frinton Laboratories FR0322 (25G)
Encompass
Salicylaldazine; Yellow Powder; 25g; M.W. 240.26; L.F. C 14 H 12 N2O2; M.P. 216 deg. to 217 deg.C; CAS: 959-36-4
Catalog No. NC9469604