Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Salicylaldazine, MP Biomedicals

Catalog No. ICN218090 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN218090 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN218090 Supplier MP Biomedicals, Inc Supplier No. 0521809080
Add to Cart
Add to Cart

Salicylaldazine, MP Biomedicals, Quantity: 100g, Molecular Weight: 240.262, CAS: 959-36-4, InChI Key: SPEXYYIULCBQJR-UHFFFAOYSA-N, IUPAC Name: 6-[[2-[(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]hydrazinyl]methylidene]cyclohexa-2,4-dien-1-one, Molecular Formula: C14H12N2O2, PubChem CID: 6849893

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Salicylaldazine
CAS 959-36-4
Molecular Formula C14H12N2O2
Quantity 100 g
Synonym salicylaldehyde azine, salicylalazine, salicylaldazine, benzaldehyde, 2-hydroxy-, 2-hydroxyphenyl methylene hydrazone, 2-hydroxybenzaldehyde azine, 2-1e-e-2-2-hydroxyphenyl methylidene hydrazin-1-ylidene methyl phenol, 2,2'-1,2-hydrazinediylidenedi methylylidene diphenol, benzaldehyde, 2-hydroxy-, 2-2-hydroxyphenyl methylene hydrazone, 1,2-bis e-salicylidene hydrazine, 6-6-oxo-1-cyclohexa-2,4-dienylidene methylhydrazo methylidene-1-cyclohexa-2,4-dienone
InChI Key SPEXYYIULCBQJR-UHFFFAOYSA-N
SMILES C1=CC(=CNNC=C2C=CC=CC2=O)C(=O)C=C1
IUPAC Name 6-[[2-[(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]hydrazinyl]methylidene]cyclohexa-2,4-dien-1-one
Molecular Weight (g/mol) 240.262
PubChem CID 6849893
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.