Learn More
Medchemexpress LLC Salicylaldoxime | 94-67-7 | 99.8% | 100 G
SDP
Catalog No. 5000411056
Encompass_Preferred
Salicylaldoxime is an organic compound that serves as a reagent for the gravimetric determination and separation of various heavy metals such as copper, nickel, palladium, lead, bismuth, and zinc. Its copper complex has also been shown to exhibit anticancer activity, specifically inhibiting topoisomerase II and L1210 leukemia cell proliferation.
  • Used as a reagent for gravimetric determination and separation of heavy metals.
  • Copper complex exhibits anticancer activity.
  • Inhibits topoisomerase II relaxation activity when complexed with copper.
  • Inhibits L1210 leukemia cell proliferation.
Catalog No. 50-004-11056 Supplier Medchemexpress LLC Supplier No. HYD0216100G
Salicylaldoxime is an organic compound that serves as a reagent for the gravimetric determination and separation of various heavy metals such as copper, nickel, palladium, lead, bismuth, and zinc. Its copper complex has also been shown to exhibit anticancer activity, specifically inhibiting topoisomerase II and L1210 leukemia cell proliferation.
  • Used as a reagent for gravimetric determination and separation of heavy metals.
  • Copper complex exhibits anticancer activity.
  • Inhibits topoisomerase II relaxation activity when complexed with copper.
  • Inhibits L1210 leukemia cell proliferation.