Learn More
Medchemexpress LLC Salicylaldoxime | 94-67-7 | 99.8% | 50 G
SDP
Catalog No. 5000410978
Encompass_Preferred
Salicylaldoxime is an organic compound that serves as a reagent for the gravimetric determination and separation of various heavy metals including Copper, Nickel, Palladium, Lead, Bismuth, and Zinc. Its copper complex has also demonstrated anticancer activity.
  • Used as a reagent for gravimetric determination and separation of copper, nickel, palladium, lead, bismuth and zinc.
  • The copper complex exhibits anticancer activity.
  • Available in solid form, white to off-white color.
  • Soluble in DMSO (100 mg/mL).
Catalog No. 50-004-10978 Supplier Medchemexpress LLC Supplier No. HYD021650G
Add to Cart
Add to Cart
Salicylaldoxime is an organic compound that serves as a reagent for the gravimetric determination and separation of various heavy metals including Copper, Nickel, Palladium, Lead, Bismuth, and Zinc. Its copper complex has also demonstrated anticancer activity.
  • Used as a reagent for gravimetric determination and separation of copper, nickel, palladium, lead, bismuth and zinc.
  • The copper complex exhibits anticancer activity.
  • Available in solid form, white to off-white color.
  • Soluble in DMSO (100 mg/mL).