Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Salicylaldoxime, 98%

Catalog No. AAA1108022
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1108022 100 g
AAA1108014 25 g
AAA1108036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1108022 Supplier Thermo Scientific Chemicals Supplier No. A1108022
Only null left
Add to Cart

CAS: 94-67-7 | C7H7NO2 | 137.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-67-7
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD00002120
InChI Key GFCNBJDXQMZOOC-WAYWQWQTSA-N
Synonym salicylaldoxime, 2-hydroxybenzaldehyde oxime, salicylaldehyde oxime, 2-hydroxyimino methyl phenol, 6-hydroxyamino methylidene cyclohexa-2,4-dien-1-one, e-2-hydroxybenzaldehyde oxime, orihzizptztncu-uhfffaoysa-n, orihzizptztncu-vmpitwqzsa-n, 2-1e-hydroxyimino methyl phenol, salicylideneaminoalcohol
PubChem CID 6740756
IUPAC Name 6-[(hydroxyamino)methylidene]cyclohexa-2,4-dien-1-one
SMILES ON\C=C1\C=CC=CC1=O

Specifications

Melting Point 55°C to 59°C
Quantity 100 g
Beilstein 1859881
Merck Index 14,8327
Formula Weight 137.14
Percent Purity 98%
Chemical Name or Material Salicylaldoxime
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 202-353-8
RTECSNumber VN5775000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.