Learn More
Medchemexpress LLC Salicylamide | 65-45-2 | 99.7% | 25 G
SDP
Catalog No. 5000406583
Encompass_Preferred
Salicylamide is an inhibitor of microsomal UDP-glucuronosyltransferase. It also acts as an analgesic and anti-pyretic agent. Studies indicate that salicylamide treatment inhibits bacterial growth and affects radiosulfate levels in maternal serum and placenta, thereby affecting the incorporation of radiosulfate into fetal skeletal GAGs and calcium content of fetal limb bones.
  • Inhibitor of microsomal UDP-glucuronosyltransferase
  • Analgesic and anti-pyretic agent
  • Inhibits bacterial growth
  • Decreases radiosulfate levels in maternal serum and placenta
  • Impairs incorporation of radiosulfate into fetal skeletal GAGs
  • Decreases calcium content of fetal limb bones
Catalog No. 50-004-06583 Supplier Medchemexpress LLC Supplier No. 25GHYB0811
Salicylamide is an inhibitor of microsomal UDP-glucuronosyltransferase. It also acts as an analgesic and anti-pyretic agent. Studies indicate that salicylamide treatment inhibits bacterial growth and affects radiosulfate levels in maternal serum and placenta, thereby affecting the incorporation of radiosulfate into fetal skeletal GAGs and calcium content of fetal limb bones.
  • Inhibitor of microsomal UDP-glucuronosyltransferase
  • Analgesic and anti-pyretic agent
  • Inhibits bacterial growth
  • Decreases radiosulfate levels in maternal serum and placenta
  • Impairs incorporation of radiosulfate into fetal skeletal GAGs
  • Decreases calcium content of fetal limb bones