Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Salicylamide, ≥99%, MP Biomedicals™

Catalog No. ICN10283591 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10283591 1 kg
ICN10283525 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10283591 Supplier MP Biomedicals, Inc Supplier No. 0210283591

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 65-45-2
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD00007978
InChI Key SKZKKFZAGNVIMN-UHFFFAOYSA-N
Synonym salicylamide, o-hydroxybenzamide, benzamide, 2-hydroxy, salicylic acid amide, 2-carbamoylphenol, flarpirina, morsarinas, algamon, algiamida, allevin
PubChem CID 5147
ChEBI CHEBI:32114
IUPAC Name 2-hydroxybenzamide
SMILES NC(=O)C1=CC=CC=C1O

Specifications

Color White
Quantity 1 kg
Formula Weight 137.1
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material Salicylamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.