Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Salicylamide, 99%

Catalog No. AC229360050
Change view
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass Bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC229360050 5 g Glass Bottle
AC229362500 250 g Plastic bottle
AC229360010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC229360050 Supplier Thermo Scientific Chemicals Supplier No. 229360050
  • $51.00
    $47.65 / Each of 1

    Save $3.35

    7% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 65-45-2 | C7H7NO2 | 137.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65-45-2
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD00007978
InChI Key SKZKKFZAGNVIMN-UHFFFAOYSA-N
Synonym salicylamide, o-hydroxybenzamide, benzamide, 2-hydroxy, salicylic acid amide, 2-carbamoylphenol, flarpirina, morsarinas, algamon, algiamida, allevin
PubChem CID 5147
ChEBI CHEBI:32114
IUPAC Name 2-hydroxybenzamide
SMILES NC(=O)C1=CC=CC=C1O

Specifications

Melting Point 139.0°C to 142.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (on anhydrous substance)
Packaging Glass Bottle
Linear Formula C6H4(OH)CONH2
Quantity 5 g
Beilstein 10, 87
Merck Index 15, 8465
Solubility Information Solubility in water: soluble in hot water. Other solubilities: 0.2% in water (30°C), 10.0% in propylene glycol (5°C), soluble in alcohol, chloroform and ether, slightly soluble in carbon tetrachloride
Formula Weight 137.14
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Salicylamide, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-609-3
RTECSNumber VN6475000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.