Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Salicylanilide 98.0+%, TCI America™
SDP

Catalog No. S000725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S000725G Supplier TCI America Supplier No. S000725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 87-17-2
Molecular Formula C13H11NO2
Molecular Weight (g/mol) 213.24
MDL Number MFCD00002212
InChI Key WKEDVNSFRWHDNR-UHFFFAOYSA-N
Synonym salicylanilide, n-phenylsalicylamide, salinide, salicylanilid, salifebrin, salinidol, ansadol, salicylic acid anilide, salnide, benzamide, 2-hydroxy-n-phenyl
PubChem CID 6872
ChEBI CHEBI:239133
IUPAC Name 2-hydroxy-N-phenylbenzamide
SMILES OC1=CC=CC=C1C(=O)NC1=CC=CC=C1

Specifications

Melting Point 137°C
Quantity 25 g
Formula Weight 213.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Salicylanilide
EINECSNumber (3)-1551
RTECSNumber VN7850000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.