Learn More
Hach Company Salicylate/Nitroprusside Reagent, 500 mL for TKN Analysis (FIA Compatible)
Catalog No. NC2009492 Shop All Hach Company Products
Encompass
Prepared salicylate/nitroprusside reagent formulated for Total Kjeldahl Nitrogen (TKN) analysis and optimized for use with flow injection analysis (FIA) systems.
  • Contains 500 mL of ready-to-use liquid reagent.
  • Used for determination of total Kjeldahl nitrogen (TKN) via salicylate/nitroprusside chemistry.
  • Compatible with flow injection analysis (FIA) systems.
  • Unopened shelf life of six months; follow method recommendations after opening.
  • Reduces reagent preparation time and supports consistent analytical results.
Catalog No. NC2009492 Supplier Hach Company Supplier No. 52010
Prepared salicylate/nitroprusside reagent formulated for Total Kjeldahl Nitrogen (TKN) analysis and optimized for use with flow injection analysis (FIA) systems.
  • Contains 500 mL of ready-to-use liquid reagent.
  • Used for determination of total Kjeldahl nitrogen (TKN) via salicylate/nitroprusside chemistry.
  • Compatible with flow injection analysis (FIA) systems.
  • Unopened shelf life of six months; follow method recommendations after opening.
  • Reduces reagent preparation time and supports consistent analytical results.