Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Salicylate/Nitroprusside Solution, EPA for Kjeldahl Nitrogen Determination, Ricca Chemical
SDP

Catalog No. 755332 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
7553-32 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 7553-32 Supplier Ricca Chemical Company Supplier No. 755332
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Salicylate / Nitroprusside Solution
CAS Min % 0.03, 12.79
Color Orange/Pink
CAS Max % 0.03, 13.31
CAS 13755-38-9, 54-21-7, 7732-18-5
Physical Form Liquid
Quantity 1 L
Packaging Amber Glass Bottle
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.