Learn More
Cayman Chemical Salicylic Acid, Purity: ≥98%, CAS: 69-72-7, 100 mg
SDP
Catalog No. 502947689
Encompass_Preferred
An analytical reference standard categorized as COX inhibitor metabolite; a metabolite of aspirin; has been found as an adulterant in seized heroin samples; has been found as an adulterant in milk; intended for research and forensic applications
Catalog No. 50-294-7689 Supplier Cayman Chemical Supplier No. 32991100 MG
An analytical reference standard categorized as COX inhibitor metabolite; a metabolite of aspirin; has been found as an adulterant in seized heroin samples; has been found as an adulterant in milk; intended for research and forensic applications