Learn More
Medchemexpress LLC Salicylic acid-13C6 (2-hydroxybenzoic acid-13C6) | 1189678-81-6 | 97.0% | 1 MG
SDP
Catalog No. 5000406937
Encompass_Preferred
Salicylic acid-13C6 is a 13C-labeled version of salicylic acid, which is a precursor and metabolite of aspirin. It can inhibit cyclo-oxygenase-2 (COX-2) activity and is intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated into drug molecules for quantitation during drug development.
  • Deuteration may affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-06937 Supplier Medchemexpress LLC Supplier No. HYB0167S11MG
Salicylic acid-13C6 is a 13C-labeled version of salicylic acid, which is a precursor and metabolite of aspirin. It can inhibit cyclo-oxygenase-2 (COX-2) activity and is intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated into drug molecules for quantitation during drug development.
  • Deuteration may affect the pharmacokinetic and metabolic profiles of drugs.