Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Solutions Salicylic Acid 15% w/v in Methanol (4L)
SDP

Catalog No. NC9683673
Encompass_Preferred

Salicylic Acid 15% w/v in Methanol (4L)

Catalog No. NC9683673 Supplier Aqua Solutions Supplier No. DC814275
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Salicylic Acid 15% w/v in Methanol (4L)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.