Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Salicylic acid 2-O-β-D-glucoside | 10366-91-3 | MFCD06203147 | >95.0% | 300.26 | C13H16O8 | 25 MG
SDP

Catalog No. 5000284875
Encompass_Preferred

Salicylic acid 2-O-β-D-glucoside is the predominant glycosylated metabolite of salicylic acid and is used as a reference compound in metabolic and enzymatic studies. The material is intended for research use and has specified storage conditions for powder and solution forms.

  • Predominant glycosylated metabolite of salicylic acid.
  • Suitable as a reference standard in metabolic and enzymatic assays.
  • CAS number 10366-91-3 and molecular weight 300.26 g/mol.
  • Storage: powder at -20°C; in solvent at -80°C (up to 6 months) or -20°C (short term).
  • Supplied in milligram-scale research pack sizes for laboratory use.

Catalog No. 50-002-84875 Supplier Medchemexpress LLC Supplier No. HYW58595225MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Salicylic acid 2-O-β-D-glucoside is the predominant glycosylated metabolite of salicylic acid and is used as a reference compound in metabolic and enzymatic studies. The material is intended for research use and has specified storage conditions for powder and solution forms.

  • Predominant glycosylated metabolite of salicylic acid.
  • Suitable as a reference standard in metabolic and enzymatic assays.
  • CAS number 10366-91-3 and molecular weight 300.26 g/mol.
  • Storage: powder at -20°C; in solvent at -80°C (up to 6 months) or -20°C (short term).
  • Supplied in milligram-scale research pack sizes for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.