Learn More
Medchemexpress LLC Salicylic acid 2-O-β-D-glucoside | 10366-91-3 | MFCD06203147 | 96.2% | 300.26 g·mol⁻¹ | C13H16O8 | 10 MG
SDP
Catalog No. 5000284626
Encompass_Preferred
Salicylic acid 2-O-β-D-glucoside is the β-D-glucoside conjugate of salicylic acid and the predominant glycosylated metabolite. Supplied as a research-grade chemical reference, it is used in metabolic profiling, enzymology, and analytical method development. Recommended storage: powder at -20°C; in solvent at -80°C for long-term storage.
  • Metabolite standard for metabolic and enzymology studies.
  • Reported high purity suitable for analytical work.
  • Molecular weight 300.26 g·mol⁻¹; formula C13H16O8.
  • Stable when stored under recommended temperature conditions.
  • Provided in small milligram quantities for reference use.
Catalog No. 50-002-84626 Supplier Medchemexpress LLC Supplier No. HYW58595210MG
Salicylic acid 2-O-β-D-glucoside is the β-D-glucoside conjugate of salicylic acid and the predominant glycosylated metabolite. Supplied as a research-grade chemical reference, it is used in metabolic profiling, enzymology, and analytical method development. Recommended storage: powder at -20°C; in solvent at -80°C for long-term storage.
  • Metabolite standard for metabolic and enzymology studies.
  • Reported high purity suitable for analytical work.
  • Molecular weight 300.26 g·mol⁻¹; formula C13H16O8.
  • Stable when stored under recommended temperature conditions.
  • Provided in small milligram quantities for reference use.