Learn More
Medchemexpress LLC Salicylic acid 2-O-β-D-glucoside | 10366-91-3 | MFCD06203147 | 96.2% | 300.26 g·mol⁻¹ | C13H16O8 | 5 MG
SDP
Catalog No. 5000284761
Encompass_Preferred
Salicylic acid 2-O-β-D-glucoside is the predominant glycosylated metabolite of salicylic acid used as a research reagent and reference standard in metabolic and analytical studies. Supplied as a powder for laboratory use, it is commonly used in metabolite profiling, assay development, and analytical method validation.
  • Predominant glycosylated metabolite of salicylic acid.
  • High purity, typically reported around 96.2%.
  • Molecular formula C13H16O8; molecular weight 300.26 g·mol⁻¹.
  • Available in small milligram-scale packages for laboratory use.
  • Stable as a powder; store at -20 °C for long-term storage.
Catalog No. 50-002-84761 Supplier Medchemexpress LLC Supplier No. HYW5859525MG
Salicylic acid 2-O-β-D-glucoside is the predominant glycosylated metabolite of salicylic acid used as a research reagent and reference standard in metabolic and analytical studies. Supplied as a powder for laboratory use, it is commonly used in metabolite profiling, assay development, and analytical method validation.
  • Predominant glycosylated metabolite of salicylic acid.
  • High purity, typically reported around 96.2%.
  • Molecular formula C13H16O8; molecular weight 300.26 g·mol⁻¹.
  • Available in small milligram-scale packages for laboratory use.
  • Stable as a powder; store at -20 °C for long-term storage.