Learn More
Medchemexpress LLC Salicylic acid 2-O-β-D-glucoside | 10366-91-3 | MFCD06203147 | 96.2% | 300.26 g/mol | C13H16O8 | 50 MG
SDP
Catalog No. 5000284760
Encompass_Preferred
Salicylic acid 2-O-β-D-glucoside is the predominant glycosylated metabolite of salicylic acid, used as a research chemical and reference standard in biochemical, metabolic, and plant signaling studies. It is typically supplied as a solid powder for analytical and preparative applications.
  • Glycosylated salicylic acid metabolite for metabolic studies
  • Solid powder form suitable for analytical assays
  • Molecular formula C13H16O8; molecular weight ≈ 300.26 g/mol
  • Available in small research pack sizes on the milligram scale
  • Reported purity approximately 96.2% based on supplier listings
Catalog No. 50-002-84760 Supplier Medchemexpress LLC Supplier No. HYW58595250MG
Salicylic acid 2-O-β-D-glucoside is the predominant glycosylated metabolite of salicylic acid, used as a research chemical and reference standard in biochemical, metabolic, and plant signaling studies. It is typically supplied as a solid powder for analytical and preparative applications.
  • Glycosylated salicylic acid metabolite for metabolic studies
  • Solid powder form suitable for analytical assays
  • Molecular formula C13H16O8; molecular weight ≈ 300.26 g/mol
  • Available in small research pack sizes on the milligram scale
  • Reported purity approximately 96.2% based on supplier listings