Learn More
Cayman Chemical Salicylic Acid, Purity: ≥98%, CAS: 69-72-7, 50 mg
SDP
Catalog No. 502951042
Encompass_Preferred
An analytical reference standard categorized as COX inhibitor metabolite; a metabolite of aspirin; has been found as an adulterant in seized heroin samples; has been found as an adulterant in milk; intended for research and forensic applications
This item is not returnable. View return policy
Catalog No. 50-295-1042 Supplier Cayman Chemical Supplier No. 3299150 MG
An analytical reference standard categorized as COX inhibitor metabolite; a metabolite of aspirin; has been found as an adulterant in seized heroin samples; has been found as an adulterant in milk; intended for research and forensic applications