Learn More
Apexbio Technology LLC Salicylic acid 69-72-7 50g
SDP
Catalog No. 5000090550
Encompass_Preferred
Salicylic acid (CAS 69-72-7) is a plant-derived phenolic compound originally isolated from willow bark It acts as an inhibitor of cyclooxygenase (COX) enzymes thereby modulating prostaglandin synthesis and reducing inflammatory signaling This mechanism underpins its widespread use in research focused on inflammation pain and related cellular pathways Salicylic acid serves as a molecular tool in studies investigating the regulation of COX activity prostaglandin-mediated responses and the development of anti-inflammatory therapeutics
Catalog No. 50-000-90550 Supplier Apexbio Technology LLC Supplier No. B109250000
Salicylic acid (CAS 69-72-7) is a plant-derived phenolic compound originally isolated from willow bark It acts as an inhibitor of cyclooxygenase (COX) enzymes thereby modulating prostaglandin synthesis and reducing inflammatory signaling This mechanism underpins its widespread use in research focused on inflammation pain and related cellular pathways Salicylic acid serves as a molecular tool in studies investigating the regulation of COX activity prostaglandin-mediated responses and the development of anti-inflammatory therapeutics