Learn More
Medchemexpress LLC Salicylic acid | 69-72-7 | 98.4% | 50 G
SDP
Catalog No. 5000406939
Encompass_Preferred
Salicylic acid (2-Hydroxybenzoic acid) is an inhibitor of cyclo-oxygenase-2 (COX-2) activity that works independently of transcription factor (NF-κB) activation. It also possesses blood-brain barrier permeability. It inhibits prostaglandin E2 release with an IC50 of 5 μg/mL and acutely causes a concentration-dependent inhibition of COX-2 activity.
  • Inhibits prostaglandin E2 release with an IC50 of 5 μg/mL
  • Acutely causes a concentration-dependent inhibition of COX-2 activity
  • Reduces both fasting and postprandial plasma glucose levels
  • Shows a trend to reduce plasma triglyceride levels
  • Significantly reduces 11β-HSD1 mRNA in omental adipose tissue
  • Reduces 11β-HSD1 enzyme activity in mesenteric adipose
Catalog No. 50-004-06939 Supplier Medchemexpress LLC Supplier No. HYB016750G
Salicylic acid (2-Hydroxybenzoic acid) is an inhibitor of cyclo-oxygenase-2 (COX-2) activity that works independently of transcription factor (NF-κB) activation. It also possesses blood-brain barrier permeability. It inhibits prostaglandin E2 release with an IC50 of 5 μg/mL and acutely causes a concentration-dependent inhibition of COX-2 activity.
  • Inhibits prostaglandin E2 release with an IC50 of 5 μg/mL
  • Acutely causes a concentration-dependent inhibition of COX-2 activity
  • Reduces both fasting and postprandial plasma glucose levels
  • Shows a trend to reduce plasma triglyceride levels
  • Significantly reduces 11β-HSD1 mRNA in omental adipose tissue
  • Reduces 11β-HSD1 enzyme activity in mesenteric adipose