Learn More
Medchemexpress LLC Salicylic acid | 69-72-7 | MFCD00002439 | 98.4% | 10 G
SDP
Catalog No. 5000406453
Encompass_Preferred
Salicylic acid (2-Hydroxybenzoic acid) is an inhibitor of cyclo-oxygenase-2 (COX-2) activity. It functions independently of transcription factor (NF-κB) activation and exhibits blood-brain barrier permeability.
  • Inhibits cyclo-oxygenase-2 (COX-2) activity
  • Functions independently of transcription factor (NF-κB) activation
  • Exhibits blood-brain barrier permeability
Catalog No. 50-004-06453 Supplier Medchemexpress LLC Supplier No. HYB016710G
Salicylic acid (2-Hydroxybenzoic acid) is an inhibitor of cyclo-oxygenase-2 (COX-2) activity. It functions independently of transcription factor (NF-κB) activation and exhibits blood-brain barrier permeability.
  • Inhibits cyclo-oxygenase-2 (COX-2) activity
  • Functions independently of transcription factor (NF-κB) activation
  • Exhibits blood-brain barrier permeability