Learn More
Selleck Chemical LLC Salicylic acid S4539-100mg
SDP
Catalog No. 503110434
Encompass_Preferred
Salicylic acid (2-Hydroxybenzoic acid) is a beta hydroxy acid that occurs as a natural compound in plants which is an inhibitor of ethylene biosynthesis and cyclooxygenase activity Salicylic acid can be used to induce animal models of Gastric and Intestinal Ulcers
Catalog No. 50-311-0434 Supplier Selleck Chemical LLC Supplier No. S4539100MG
Salicylic acid (2-Hydroxybenzoic acid) is a beta hydroxy acid that occurs as a natural compound in plants which is an inhibitor of ethylene biosynthesis and cyclooxygenase activity Salicylic acid can be used to induce animal models of Gastric and Intestinal Ulcers