Learn More
Selleck Chemical LLC Salicylic acid S4539-500mg
SDP
Catalog No. 503111493
Encompass_Preferred
Salicylic acid (2-Hydroxybenzoic acid) is a beta hydroxy acid that occurs as a natural compound in plants which is an inhibitor of ethylene biosynthesis and cyclooxygenase activity Salicylic acid can be used to induce animal models of Gastric and Intestinal Ulcers
Catalog No. 50-311-1493 Supplier Selleck Chemical LLC Supplier No. S4539500MG
Salicylic acid (2-Hydroxybenzoic acid) is a beta hydroxy acid that occurs as a natural compound in plants which is an inhibitor of ethylene biosynthesis and cyclooxygenase activity Salicylic acid can be used to induce animal models of Gastric and Intestinal Ulcers