Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Salicylic Acid (Crystalline/USP), Fisher Chemical

Catalog No. A275500 Shop All Fisherbrand Products
Change view
Click to view available options
Poly Bottle; 500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
A275-500 Poly Bottle; 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A275-500

missing translation for 'validMainframeMsg'

Specifications

pH 2.4
Melting Point 158°C
CAS 69-72-7
Boiling Point 211°C
Color White
Molecular Formula C7H6O3
Linear Formula 2-HOC6H4COOH
MDL Number MFCD00002439
Synonym salicylic acid, o-hydroxybenzoic acid, 2-carboxyphenol, o-carboxyphenol, rutranex, salonil, retarder w, keralyt, duoplant, freezone
InChI Key YGSDEFSMJLZEOE-UHFFFAOYSA-N
SMILES OC(=O)C1=CC=CC=C1O
IUPAC Name 2-hydroxybenzoic acid
Molecular Weight (g/mol) 138.12
PubChem CID 338
ChEBI CHEBI:16914
Formula Weight 138.12
Percent Purity 99.5 to 101.0% (USP)
Grade USP
Quantity 500 g
Ignition Residue 0.05% max. (USP)
Loss on Drying 0.5% max. (USP)
Packaging Poly Bottle
Physical Form Solid
Vapor Pressure 0.3mbar at 95°C
Chemical Name or Material Salicylic Acid
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Category SIGNAL WORD: DANGER!
Precautionary Statement Emergency Overview
Harmful if swallowed. Irritating to eyes, respiratory system and skin.
NFPA
Health: 2
Flammability: 1
Instability: 0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.