Learn More
Cayman Chemical Salicylic Acid-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 78646-17-0, 10 mg
SDP
Catalog No. 502913952
Encompass_Preferred
An internal standard for the quantification of salicylic acid by GC- or LC-MS
Catalog No. 50-291-3952 Supplier Cayman Chemical Supplier No. 3541810 MG
An internal standard for the quantification of salicylic acid by GC- or LC-MS