Learn More
Cayman Chemical Salicylic Acid-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 78646-17-0, 25 mg
SDP
Catalog No. 502909441
Encompass_Preferred
An internal standard for the quantification of salicylic acid by GC- or LC-MS
Catalog No. 50-290-9441 Supplier Cayman Chemical Supplier No. 3541825 MG
An internal standard for the quantification of salicylic acid by GC- or LC-MS