Learn More
Medchemexpress LLC Salicylic acid-d6 (2-hydroxybenzoic acid-d6) | 285979-87-5 | 99.8% | 1 MG
Supplier: Medchemexpress LLC HYB0167S1MG
Salicylic acid-d6 is a deuterium labeled Salicylic acid that inhibits cyclo-oxygenase-2 (COX-2) activity independently of transcription factor (NF-κB) activation. It is for research use only.
- Can be used as a tracer
- Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS