Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Salicylic Acid Lithium Salt, TCI America™
SDP

Catalog No. S038425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S038425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S038425G Supplier TCI America Supplier No. S038425G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 552-38-5
Molecular Formula C7H5LiO3
Molecular Weight (g/mol) 144.054
MDL Number MFCD00045813
InChI Key PSBOOKLOXQFNPZ-UHFFFAOYSA-M
Synonym lithium salicylate, unii-93f1sp6qin, salicylic acid lithium salt, dilithium salicylate, 93f1sp6qin, lithium 2-hydroxybenzoate, lithium 1+ ion salicylate, lithiumsalicylate, lithiumsalicylat, salicylicacidlithiumsalt
PubChem CID 23663621
IUPAC Name lithium;2-hydroxybenzoate
SMILES [Li+].C1=CC=C(C(=C1)C(=O)[O-])O

Specifications

Quantity 25 g
Formula Weight 144.05
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Salicylic Acid Lithium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.