Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Salicylic Acid, Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111015269 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5269 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5269 Supplier MilliporeSigma Supelco Supplier No. PHR10131G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Salicylic acid is an important material in pharmaceutical industry, obtained via the decomposition of acetylsalicylic acid. Salicylic acid is widely used in food industry for producing dyes and artificial scents. It also shows therapeutic potential in protecting the skin from sun damage and hence finds applications in cosmetics and medicine.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
TRUSTED_SUSTAINABILITY

Specifications

CAS 69-72-7
Vapor Pressure 1 mmHg (114°C)
Vapor Density 4.8 (vs. air)
Physical Form Neat
Linear Formula 2-(HO)C6H4CO2H
MDL Number MFCD00002439
Melting Point 158°C to 161°C
Boiling Point 211°C
Synonym 2-Hydroxybenzoic acid
Quantity 1 g
Formula Weight 138.12
Grade Certified Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.