Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Salicylic acid (phenyl-13C6), ≥97.5% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111012711 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2711 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2711 Supplier MilliporeSigma Supelco Supplier No. 438255MG
Add to Cart
Add to Cart
Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

CAS 1189678-81-6
Flash Point 157°C
Physical Form Neat
Linear Formula 2-(HO)13C6H4 CO2 H
MDL Number MFCD20257120
Melting Point 158°C to 161°C
Synonym 2-Hydroxybenzoic acid (phenyl-13C6)
Type Analytical Standard
Quantity 5 mg
Formula Weight 144.08
Percent Purity ≥97.5% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.