Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Salicylic acid (Standard) | 69-72-7 | 98.0% | 50 MG
SDP

Catalog No. 5000407169
Encompass_Preferred

Salicylic acid (Standard) is an analytical standard intended for research and analytical applications. It inhibits cyclo-oxygenase-2 (COX-2) activity independently of transcription factor (NF-κB) activation. This product is commonly used in qualitative, quantitative, and methodological research experiments.

  • Analytical standard for research and analytical applications
  • Inhibits cyclo-oxygenase-2 (COX-2) activity
  • Independently inhibits transcription factor (NF-κB) activation
  • Commonly used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS
  • Molecular weight: 138.12
  • Formula: C7H6O3
  • Appearance: white to off-white solid
  • Initial source from plants
  • Endogenous metabolite

Catalog No. 50-004-07169 Supplier Medchemexpress LLC Supplier No. HYB0167R50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Salicylic acid (Standard) is an analytical standard intended for research and analytical applications. It inhibits cyclo-oxygenase-2 (COX-2) activity independently of transcription factor (NF-κB) activation. This product is commonly used in qualitative, quantitative, and methodological research experiments.

  • Analytical standard for research and analytical applications
  • Inhibits cyclo-oxygenase-2 (COX-2) activity
  • Independently inhibits transcription factor (NF-κB) activation
  • Commonly used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS
  • Molecular weight: 138.12
  • Formula: C7H6O3
  • Appearance: white to off-white solid
  • Initial source from plants
  • Endogenous metabolite

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.