Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Salicylidene salicylhydrazide, 97%, Thermo Scientific™

Catalog No. AAL0823514
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0823514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0823514 Supplier Thermo Scientific Chemicals Supplier No. L0823514

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3232-36-8
Molecular Formula C14H12N2O3
Molecular Weight (g/mol) 256.26
MDL Number MFCD00043494
InChI Key NCCJLHWEUIAECU-MDZDMXLPSA-N
Synonym salicylidene salicylhydrazide, benzoic acid, 2-hydroxy-, 2-hydroxyphenyl methylene hydrazide, benzoic acid, 2-hydroxy-, 2-2-hydroxyphenyl methylene hydrazide, 2-hydroxy-n'-1e-6-oxocyclohexa-2,4-dien-1-ylidene methyl benzohydrazide, scs, 2-hydroxybenzylidene salicylohydrazide, tocris-2143, 2-hydroxy-n'-e-2-hydroxyphenyl methylidene benzohydrazide, n-1e-2-2-hydroxyphenyl-1-azavinyl 2-hydroxyphenyl carboxamide
PubChem CID 5483116
IUPAC Name 2-hydroxy-N'-[(E)-(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]benzohydrazide
SMILES OC1=CC=CC=C1C(=O)NN\C=C1/C=CC=CC1=O

Specifications

Melting Point 280°C to 284°C
Quantity 25 g
Beilstein 2944701
Formula Weight 256.26
Percent Purity 97%
Chemical Name or Material Salicylidene salicylhydrazide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 221-773-2
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.