Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Salicylideneaniline, 98%

Catalog No. p-7028174
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2533209 10 g
AAB2533218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2533209 Supplier Thermo Scientific Chemicals Supplier No. B2533209
Only null left
Add to Cart

CAS: 779-84-0 | C13H11NO | 197.237 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 779-84-0
Molecular Formula C13H11NO
Molecular Weight (g/mol) 197.237
MDL Number MFCD00020066
InChI Key GVHXCXKLYZOBKS-UHFFFAOYSA-N
Synonym n-salicylideneaniline, phenol, 2-e-phenylimino methyl, n-salicylalaniline, 2-hydroxybenzalaniline, o-hydroxy benzylidene aniline, acmc-20m6ov, acmc-1bijs, n-o-hydroxybenzylidene aniline, 2-e-phenyliminomethyl phenol, phenol,2-phenylimino methyl
PubChem CID 6740751
IUPAC Name 6-(anilinomethylidene)cyclohexa-2,4-dien-1-one
SMILES C1=CC=C(C=C1)NC=C2C=CC=CC2=O

Specifications

Melting Point 48°C to 52°C
Boiling Point 151°C (2 mmHg)
Quantity 10 g
Beilstein 1367639
Formula Weight 197.24
Percent Purity 98%
Chemical Name or Material Salicylideneaniline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.