Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Salicyluric acid | 487-54-7 | 99.8% | 195.17 | 250 MG
SDP

Catalog No. 5000439510
Encompass_Preferred

Salicyluric acid is an endogenous metabolite, exclusively for research purposes and not intended for patient use. This product is provided as a solid, white to off-white in appearance, with a molecular weight of 195.17. It is classified under phenols, monophenols, ketones, aldehydes, and acids, targeting both microbial and human endogenous metabolites.

  • Molecular weight: 195.17
  • Formula: C9H9NO4
  • CAS no.: 487-54-7
  • Appearance: Solid
  • Color: White to off-white
  • Structure classification: Phenols, monophenols, ketones, aldehydes, acids
  • Initial source: Endogenous metabolite
  • Target: Endogenous metabolite
  • Pathway: Metabolic enzyme/protease
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-39510 Supplier Medchemexpress LLC Supplier No. 250MGHY113295
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Salicyluric acid is an endogenous metabolite, exclusively for research purposes and not intended for patient use. This product is provided as a solid, white to off-white in appearance, with a molecular weight of 195.17. It is classified under phenols, monophenols, ketones, aldehydes, and acids, targeting both microbial and human endogenous metabolites.

  • Molecular weight: 195.17
  • Formula: C9H9NO4
  • CAS no.: 487-54-7
  • Appearance: Solid
  • Color: White to off-white
  • Structure classification: Phenols, monophenols, ketones, aldehydes, acids
  • Initial source: Endogenous metabolite
  • Target: Endogenous metabolite
  • Pathway: Metabolic enzyme/protease
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.