Learn More
Cayman Chemical Salidroside, Purity: ≥97%, CAS: 10338-51-9, 10 mg
SDP
Catalog No. 502953528
Encompass_Preferred
A glycoside; scavenges DPPH and ABTS radicals (EC50s = 81.54 and 30.94 µg/ml, respectively, in cell-free assays); inhibits apoptosis and production of ROS induced by Aβ (23-35) in SH-SY5Y neuroblastoma cells at 50 and 100 µM; decreases infarct volume by 17.9% in a rat model of focal cerebral ischemia-reperfusion injury induced by MCAO at 12 mg/kg; increases survival and reduces plasma ALT, AST, TNF-α, and MDA levels in a mouse model of acetaminophen-induced liver injury
Catalog No. 50-295-3528 Supplier Cayman Chemical Supplier No. 1362810 MG
A glycoside; scavenges DPPH and ABTS radicals (EC50s = 81.54 and 30.94 µg/ml, respectively, in cell-free assays); inhibits apoptosis and production of ROS induced by Aβ (23-35) in SH-SY5Y neuroblastoma cells at 50 and 100 µM; decreases infarct volume by 17.9% in a rat model of focal cerebral ischemia-reperfusion injury induced by MCAO at 12 mg/kg; increases survival and reduces plasma ALT, AST, TNF-α, and MDA levels in a mouse model of acetaminophen-induced liver injury