Learn More
Apexbio Technology LLC Salidroside 10338-51-9 20mg
SDP
Catalog No. 502240241
Encompass_Preferred
Salidroside (CAS 10338-51-9) is a naturally occurring phenylpropanoid glycoside isolated primarily from Rhodiola species It exerts multiple biological effects through modulation of cellular pathways related to oxidative stress inflammation and apoptosis including activation of the PI3K/Akt signaling pathway and inhibition of pro-inflammatory mediators Salidroside has been shown to protect cells from oxidative damage modulate mitochondrial function and suppress inflammatory responses in various in vitro and in vivo models This compound is widely utilized in biomedical research investigating neuroprotection cardioprotection and potential therapeutic strategies targeting metabolic and neurodegenerative disorders
Catalog No. 50-224-0241 Supplier Apexbio Technology LLC Supplier No. N178920
Salidroside (CAS 10338-51-9) is a naturally occurring phenylpropanoid glycoside isolated primarily from Rhodiola species It exerts multiple biological effects through modulation of cellular pathways related to oxidative stress inflammation and apoptosis including activation of the PI3K/Akt signaling pathway and inhibition of pro-inflammatory mediators Salidroside has been shown to protect cells from oxidative damage modulate mitochondrial function and suppress inflammatory responses in various in vitro and in vivo models This compound is widely utilized in biomedical research investigating neuroprotection cardioprotection and potential therapeutic strategies targeting metabolic and neurodegenerative disorders