Learn More
AdipoGen Salidroside
SDP
Catalog No. 502560064
Encompass_Preferred
Chemical. CAS 10338-51-9. Formula C14H20O7. MW 300.3. Synthetic. Glycoside isolated from plants, most commonly from members of the genus Rhodiola, with many biological activities. Shown to have neuroprotective, antioxidant, antiviral, antidepressant, cardioprotective, anti-inflammatory and anticancer activity. Ameliorates insulin resistance.
Catalog No. 50-256-0064 Supplier AdipoGen Supplier No. CDXS0092M100
Chemical. CAS 10338-51-9. Formula C14H20O7. MW 300.3. Synthetic. Glycoside isolated from plants, most commonly from members of the genus Rhodiola, with many biological activities. Shown to have neuroprotective, antioxidant, antiviral, antidepressant, cardioprotective, anti-inflammatory and anticancer activity. Ameliorates insulin resistance.