Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Salidroside, ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111012343 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2343 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2343 Supplier MilliporeSigma Supelco Supplier No. 4386625MG
Only null left
Add to Cart
Add to Cart

Salidroside is one of the major phenolic glycoside extracted from Rhodiola rosea L(, which can show a broad-spectrum of pharmacological properties like anti-aging, anti-oxidant, anti-cancer and anti-inflammation.

Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 10338-51-9
Physical Form Neat
Linear Formula C14H20O7
MDL Number MFCD00210553
Synonym 2-(4-Hydroxyphenyl)ethyl beta-D-glucopyranoside; p-Hydroxyphenethyl glucopyranoside; Rhodioloside; Rhodioloside; Rhodosin; Tyrosol a-(beta-D-glucopyranoside)
Type Analytical Standard
Quantity 25 mg
Formula Weight 300.30
Percent Purity ≥98.0% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.