Learn More
Alkali Scientific Saline 0.85% 100mL Bottle, 1/Pk
SDP
Catalog No. 5000662939
Encompass_Preferred
Saline 0.85% is a sterile isotonic sodium chloride solution supplied in a convenient 100mL bottle for general laboratory use. The balanced salt concentration helps maintain the structural integrity of microorganisms by preventing osmotic stress during washing, suspension, or dilution procedures. This bottle offers a versatile intermediate volume that is suitable for routine microbiological tasks without requiring large bulk containers. Laboratories commonly use it for rinsing filtration equipment, preparing microbial suspensions, and creating customized dilution volumes. Because the solution contains no nutrients, it provides a neutral environment that preserves cell viability without stimulating microbial growth during testing procedures.
Catalog No. 50-006-62939 Supplier Alkali Scientific Supplier No. SMSB00
Saline 0.85% is a sterile isotonic sodium chloride solution supplied in a convenient 100mL bottle for general laboratory use. The balanced salt concentration helps maintain the structural integrity of microorganisms by preventing osmotic stress during washing, suspension, or dilution procedures. This bottle offers a versatile intermediate volume that is suitable for routine microbiological tasks without requiring large bulk containers. Laboratories commonly use it for rinsing filtration equipment, preparing microbial suspensions, and creating customized dilution volumes. Because the solution contains no nutrients, it provides a neutral environment that preserves cell viability without stimulating microbial growth during testing procedures.