Learn More
Alkali Scientific Saline 0.85% 3mL 13x100mm Polycarbonate Screw Cap Tube, 1/Pk
SDP
Catalog No. 5000663182
Encompass_Preferred
Saline 0.85% 3mL 13x100mm Polycarbonate Screw Cap Tube, 1/Pk is a sterile isotonic saline solution designed for microbiological and clinical laboratory applications. The 0.85% sodium chloride concentration provides osmotic balance that maintains cell integrity without supplying nutrients that could promote microbial growth. This makes the solution suitable for preparing microbial suspensions, performing serial dilutions, rinsing cells, and supporting sample handling procedures. Each tube is prefilled with 3mL of sterile saline and packaged in a durable polycarbonate screw cap tube that helps prevent contamination while ensuring convenient handling. The single use format supports reliable, consistent workflows in clinical diagnostics, microbiology laboratories, and research environments.
Catalog No. 50-006-63182 Supplier Alkali Scientific Supplier No. SALT01
Add to Cart
Add to Cart
Saline 0.85% 3mL 13x100mm Polycarbonate Screw Cap Tube, 1/Pk is a sterile isotonic saline solution designed for microbiological and clinical laboratory applications. The 0.85% sodium chloride concentration provides osmotic balance that maintains cell integrity without supplying nutrients that could promote microbial growth. This makes the solution suitable for preparing microbial suspensions, performing serial dilutions, rinsing cells, and supporting sample handling procedures. Each tube is prefilled with 3mL of sterile saline and packaged in a durable polycarbonate screw cap tube that helps prevent contamination while ensuring convenient handling. The single use format supports reliable, consistent workflows in clinical diagnostics, microbiology laboratories, and research environments.