Learn More
Alkali Scientific Saline 0.85% 5mL Screw Cap Tube, 10/Pk
SDP
Catalog No. 5000662793
Encompass_Preferred
Saline 0.85% is a sterile isotonic sodium chloride solution designed for suspending and washing microorganisms without damaging their cellular structure. This version features a leak resistant screw cap that improves safety during vigorous mixing and transport. The secure closure allows technicians to vortex samples to create smooth microbial suspensions without risk of spills or aerosol formation. Because the solution maintains osmotic balance, it protects delicate cells from bursting or shrinking during laboratory handling. The 5mL format provides a convenient working volume for microbial suspension preparation, biochemical testing, and sample transfer procedures in microbiology laboratories where sterile and reliable diluents are required.
Catalog No. 50-006-62793 Supplier Alkali Scientific Supplier No. SMST04
Saline 0.85% is a sterile isotonic sodium chloride solution designed for suspending and washing microorganisms without damaging their cellular structure. This version features a leak resistant screw cap that improves safety during vigorous mixing and transport. The secure closure allows technicians to vortex samples to create smooth microbial suspensions without risk of spills or aerosol formation. Because the solution maintains osmotic balance, it protects delicate cells from bursting or shrinking during laboratory handling. The 5mL format provides a convenient working volume for microbial suspension preparation, biochemical testing, and sample transfer procedures in microbiology laboratories where sterile and reliable diluents are required.