Learn More
Alkali Scientific Saline 0.85% 9.9mL Screw Cap Tube, 10/Pk
SDP
Catalog No. 5000662835
Encompass_Preferred
Saline 0.85% is a sterile isotonic sodium chloride solution used to maintain cell viability during laboratory procedures. The solution provides osmotic stability that prevents microbial cells from lysing or shrinking during dilution and mixing. This specialized 9.9mL screw cap tube is calibrated to simplify high factor dilution protocols. When 0.1mL of a sample is added, the tube produces an immediate 1:100 dilution in a single step. This eliminates additional dilution stages and improves workflow efficiency in microbiology laboratories. The secure screw cap allows safe vortex mixing and transport, making the tube suitable for microbial enumeration, sample preparation, and routine dilution procedures.
Catalog No. 50-006-62835 Supplier Alkali Scientific Supplier No. SMST06
Saline 0.85% is a sterile isotonic sodium chloride solution used to maintain cell viability during laboratory procedures. The solution provides osmotic stability that prevents microbial cells from lysing or shrinking during dilution and mixing. This specialized 9.9mL screw cap tube is calibrated to simplify high factor dilution protocols. When 0.1mL of a sample is added, the tube produces an immediate 1:100 dilution in a single step. This eliminates additional dilution stages and improves workflow efficiency in microbiology laboratories. The secure screw cap allows safe vortex mixing and transport, making the tube suitable for microbial enumeration, sample preparation, and routine dilution procedures.