Learn More
Chrono Log Corporation Saline (0.9%), 15mL
SDP
Catalog No. NC1634306
Encompass
This item has a minimum qty of 4 per supplier requirements.
Sterile physiological saline for irrigation (0.85% or 0.9% w/v) for CHRONOPAR Reagent preparation and for dilution of the Whole Blood specimen.
Catalog No. NC1634306 Supplier Chrono Log Corporation Supplier No. 397
This item has a minimum qty of 4 per supplier requirements.
Sterile physiological saline for irrigation (0.85% or 0.9% w/v) for CHRONOPAR Reagent preparation and for dilution of the Whole Blood specimen.