Learn More
Alkali Scientific Saline Buffered Peptone Water 90mL Bottle, 1/Pk
SDP
Catalog No. 5000662861
Encompass_Preferred
Saline Buffered Peptone Water is a non selective diluent and pre enrichment medium widely used in food safety and environmental microbiology testing. The formulation contains peptone to provide mild nutritional support that helps recover stressed or injured bacteria, while the phosphate buffer maintains a stable neutral pH during incubation. This 90mL bottle is specifically measured to support the standard 1:10 dilution required in many regulatory testing protocols. By adding 10g of a food or environmental sample, laboratories can quickly create a properly diluted homogenate for microbiological analysis. The medium is commonly used as the first step in pathogen detection procedures before transferring samples to selective enrichment media.
Catalog No. 50-006-62861 Supplier Alkali Scientific Supplier No. SBPB00
Saline Buffered Peptone Water is a non selective diluent and pre enrichment medium widely used in food safety and environmental microbiology testing. The formulation contains peptone to provide mild nutritional support that helps recover stressed or injured bacteria, while the phosphate buffer maintains a stable neutral pH during incubation. This 90mL bottle is specifically measured to support the standard 1:10 dilution required in many regulatory testing protocols. By adding 10g of a food or environmental sample, laboratories can quickly create a properly diluted homogenate for microbiological analysis. The medium is commonly used as the first step in pathogen detection procedures before transferring samples to selective enrichment media.