Learn More
Med Vet International Saline Flush Prefilled Syringe, 10mL, J1705, Each
Catalog No. 5000582604
Encompass_Preferred
Saline Flush Prefilled Syringe. • Quicker and convenient • Sterile fluid pathway • 5ml USP 0.9% Sodium Chloride in 12ml syringe • Male luer lock with protective cap • FDA cleared • Non-pyrogenic and DEHP, PVC, Latex free • 30/box, 180/case
Catalog No. 50-005-82604 Supplier Med Vet International Supplier No. JOR1705
Saline Flush Prefilled Syringe. • Quicker and convenient • Sterile fluid pathway • 5ml USP 0.9% Sodium Chloride in 12ml syringe • Male luer lock with protective cap • FDA cleared • Non-pyrogenic and DEHP, PVC, Latex free • 30/box, 180/case