Learn More
Med Vet International Sterile Saline Solution 0.9%, 1000mL, Each
Catalog No. 502828487
Encompass_Preferred
For use in replacement therapy of sodium, chloride and water which may become depleted in many diseases in cattle, swine, sheep, goats, horses, dogs and cats. . Because this solution is isotonic with body fluids, it may also be used as a solvent or diluent for antibiotics and other pharmaceuticals and biologicals where compatible, and for washing mucous membranes and other tissue surfaces. Each 100 mL contains 0.9 gram of sodium chloride. Packaged in a hard plastic bottle that has a fold-flat integrated hook for hanging.
Catalog No. 50-282-8487 Supplier Med Vet International Supplier No. RXSALINE0.91000V
For use in replacement therapy of sodium, chloride and water which may become depleted in many diseases in cattle, swine, sheep, goats, horses, dogs and cats. . Because this solution is isotonic with body fluids, it may also be used as a solvent or diluent for antibiotics and other pharmaceuticals and biologicals where compatible, and for washing mucous membranes and other tissue surfaces. Each 100 mL contains 0.9 gram of sodium chloride. Packaged in a hard plastic bottle that has a fold-flat integrated hook for hanging.