Learn More
Thermco Products Inc Plastic hydrometer for salt brine, 1-100%, 1.0 div, 300mm
Catalog No. NC0262463
Encompass_Preferred
Made from virgin polycarbonate. Shatter proof with clarity like glass Calibrated against N.I.S.T. standards. May be certified and traceable to N.I.S.T. Approximately 300mm. Measures the saturatedsalt in aqueous solution.
Catalog No. NC0262463 Supplier Thermco Products Inc Supplier No. ACC8511PC
Made from virgin polycarbonate. Shatter proof with clarity like glass Calibrated against N.I.S.T. standards. May be certified and traceable to N.I.S.T. Approximately 300mm. Measures the saturatedsalt in aqueous solution.