Learn More
Azzota Corp Hand Held Salinity Refractometer, Measures Salt In Sea Water And Prepared Food Solution, Scale Range: 0 To 28%, Minimum Scale: 0.2%, Accuracy: 0.2%
Catalog No. 507532944
Encompass_Preferred
Hand Held Salinity Refractometer, Measure salt in sea water and prepared food solution, Scale Range: 0 to 28%, Minimum scale: 0.2%, Accuracy: 0.2%
Catalog No. 50-753-2944 Supplier Azzota Corp Supplier No. REF211
Hand Held Salinity Refractometer, Measure salt in sea water and prepared food solution, Scale Range: 0 to 28%, Minimum scale: 0.2%, Accuracy: 0.2%